2-Propanol ≥99.9%, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™

Supplier: BURDICK JACKSON
Danger

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

PP323-19 PP323-204
BJAAPP323-19EA 1530 SGD
BJAAPP323-19 BJAAPP323-204
2-Propanol ≥99.9%, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™
2-Propanol

  • Low residue
  • Purified solvent
  • ACS General Use certified

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 12.0 %(V)
Melting Pt: –88 °C
Density: 0.785 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay by GC Analysis 99.9 % min.
Carbonyl Compounds 0.002 % max.
Color (APHA) 10 max.
Refractive Index (20ºC) 1.3762 - 1.3782
Residue after Evaporation 5 mg/L max.
Solubility in Water Passes test
Titrable Acid or Base 0.1 µeq/g max.
Water by Karl Fischer Titration 0.1 % max.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR