Tetrahydrofuran ≥99.9% stabilised, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™

Supplier: BURDICK JACKSON
Danger


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

PP341-19 PP341-204
BJAAPP341-19EA 2380 SGD
BJAAPP341-19 BJAAPP341-204
Tetrahydrofuran ≥99.9% stabilised, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™
Tetrahydrofuran

  • Low residue
  • Purified solvent
  • ACS General Use certified

Formula: C₄H₈O
Boiling Pt: 11,8 %(V)
Melting Pt: –108,5 °C
Density: 0,8892 g/cm³ (20 °C)
Flash Pt: –14 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay by GC Analysis 99.9 % min.
Color (APHA) 20 max.
Peroxides (as H₂O₂) at time of packaging 2 mg/L max.
Refractive Index (20ºC) 1.4060 - 1.4080
Residue after Evaporation 0.03 mg/L max.
Water by Karl Fischer Titration 0.05 % max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR