2,2,4-Trimethylpentane ≥99.0%, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™

Supplier: BURDICK & JACKSON
Danger

PP362-204 PP362TC19
BJAAPP362-204EA 20740 SGD
BJAAPP362-204 BJAAPP362TC19
2,2,4-Trimethylpentane ≥99.0%, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™
2,2,4-Trimethylpentane

  • Low residue
  • Purified solvent
  • ACS General Use certified

Formula: (CH₃)₂CHCH₂C(CH₃)₃
Boiling Pt: 99,24 °C
Melting Pt: -107,4 °C
Density: 0,69 g/cm³ (20 °C)
Flash Pt: -8 °C
Storage Temperature: Ambient
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3,II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=559f761f651e5fcdb0cf5dd303cd79e53daf2366&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay by GC Analysis 99 % min.
Color (APHA) 10 max.
Refractive Index (20ºC) 1.3907 - 1.3923
Residue after Evaporation 5 mg/L max.
Sulfur Compounds (as S) 0.005 % max.
Water by Karl Fischer Titration 0.01 % max.
Water-Soluble Titrable Acid 0.3 µeq/g max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR