2-Benzylaminoethanol 96%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
N-Benzylethanolamine, 2-(Benzylamino)ethanol
A16866.0E
A16866.36
ALFAA16866.0EEA
0
SGD
ALFAA16866.0E
ALFAA16866.36
2-Benzylaminoethanol 96%
2-Benzylaminoethanol
Purity: 96%
CAS No.: 104-63-2
Molecular Formula: C6H5CH2NHCH2CH2OH
Formula Weight: 151.21
Flash Point: 130°(266°F)
Refractive Index: 1.5430
Melting Point: -30°
Boiling Point: 240°
Beilstein Reference: 775164
MDL No.: MFCD00002840
Formula:
C₆H₅CH₂NHCH₂CH₂OH MW: 151,21 g/mol Boiling Pt: 240 °C Melting Pt: –30 °C Density: 1,063 Flash Pt: 130 °C (266 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002840 CAS Number: 104-63-2 EINECS: 203-221-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...