5-Bromo-2-hydroxybenzyl alcohol ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Bromo-2-(hydroxymethyl)phenol, (5-Bromo-2-hydroxyphenyl)methanol

A12344.06 A12344.14
ALFAA12344.06EA 0 SGD
ALFAA12344.06 ALFAA12344.14
5-Bromo-2-hydroxybenzyl alcohol ≥98%
5-Bromo-2-hydroxybenzyl alcohol

Purity: 98+% CAS No.: 2316-64-5 Molecular Formula: C7H7BrO2 Formula Weight: 203.04 Melting Point: 108-110° Beilstein Reference: 509726

Formula: C₇H₇BrO₂
MW: 203,04 g/mol
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004618
CAS Number: 2316-64-5
EINECS: 219-026-0
Merck Index: 12,01460

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR