2-Chloro-4,5-xylenol ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Chloro-4,5-dimethylphenol, 4-Chloro-5-hydroxy-2-xylene, 6-Chloro-3,4-xylenol

L01627.03 L01627.06
ALFAL01627.03EA 0 SGD
ALFAL01627.03 ALFAL01627.06
2-Chloro-4,5-xylenol ≥98%
2-Chloro-4,5-xylenol

Purity: 98% CAS No.: 1124-04-5 Molecular Formula: C8H9ClO Formula Weight: 156.61 Melting Point: 70-72° Beilstein Reference: 2042057

Formula: C₈H₉ClO
MW: 156,61 g/mol
Melting Pt: 70…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00002173
CAS Number: 1124-04-5
EINECS: 214-386-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR