3-Fluoro-4-methoxybenzyl alcohol ≥98%

Supplier: Thermo Scientific
Warning

H32350.03 H32350.09
ALFAH32350.03EA 0 SGD
ALFAH32350.03 ALFAH32350.09
3-Fluoro-4-methoxybenzyl alcohol ≥98%
3-Fluoro-4-methoxybenzyl alcohol

Purity: 98% CAS No.: 96047-32-4 Molecular Formula: C8H9FO2 Formula Weight: 156.15 MDL No.: MFCD00070642

Formula: C₈H₉FO₂
MW: 156,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070642
CAS Number: 96047-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR