3-Fluoro-4-methylbenzyl alcohol ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(3-Fluoro-4-methylphenyl)methanol
L19170.03
L19170.MD
ALFAL19170.03EA
0
SGD
ALFAL19170.03
ALFAL19170.MD
3-Fluoro-4-methylbenzyl alcohol ≥97%
3-Fluoro-4-methylbenzyl alcohol
Purity: 97%
CAS No.: 192702-79-7
Molecular Formula: C8H9FO
Formula Weight: 140.16
Melting Point: 33-36°
Beilstein Reference: 7700178
Formula:
C₈H₉FO MW: 140,16 g/mol Melting Pt: 33…36 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631533 CAS Number: 192702-79-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...