2-Bromonicotinic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Bromo-3-pyridinecarboxylic acid, 2-Bromopyridine-3-carboxylic acid
H30239.03
H30239.06
ALFAH30239.03EA
0
SGD
ALFAH30239.03
ALFAH30239.06
2-Bromonicotinic acid ≥97%
2-Bromonicotinic acid
Purity: 97%
CAS No.: 35905-85-2
Molecular Formula: C6H4BrNO2
Formula Weight: 202.01
Melting Point: ca 254°
MDL No.: MFCD01646068
Formula:
C₆H₄BrNO₂ MW: 202,01 g/mol Melting Pt: ∼ 254 °C Storage Temperature: Ambient |
MDL Number:
MFCD01646068 CAS Number: 35905-85-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...