2-Octynoic acid ethyl ester ≥98%
Supplier: Thermo Scientific
Synonyms:
Ethyl 2-octynoate
A19784.06
A19784.14
ALFAA19784.06EA
0
SGD
ALFAA19784.06
ALFAA19784.14
2-Octynoic acid ethyl ester ≥98%
2-Octynoic acid ethyl ester
Purity: 98%
CAS No.: 10519-20-7
Molecular Formula: C10H16O2
Formula Weight: 168.24
Storage Temperature: 2°C to 8°C
Flash Point: 99°(210°F)
Refractive Index: 1.4470
Boiling Point: 85°/6mm
Beilstein Reference: 1759822
MDL No.: MFCD00015286
Formula:
C₁₀H₁₆O₂ MW: 168,24 g/mol Boiling Pt: 85 °C (6 mmHg) Density: 0,910 Flash Pt: 99 °C (210 °F) |
MDL Number:
MFCD00015286 CAS Number: 10519-20-7 EINECS: 234-058-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...