3-Iodophenylboronic acid ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Dihydroxy(3-iodophenyl)borane, (3-Iodophenyl)boronic acid, 3-Iodobenzeneboronic acid, (3-Iodophenyl)metaboric acid
H51112.03
H51112.06
ALFAH51112.03EA
59.6
SGD
ALFAH51112.03
ALFAH51112.06
3-Iodophenylboronic acid ≥97%
3-Iodophenylboronic acid
Purity: 97%
CAS No.: 221037-98-5
Molecular Formula: C6H6BIO2
Formula Weight: 247.83
MDL No.: MFCD01319015
Formula:
C₆H₆BIO₂ MW: 247,83 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01319015 CAS Number: 221037-98-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...