4-Aminonicotinic acid 98%

Supplier: Thermo Scientific
Warning

H50032.03 H50032.06
ALFAH50032.03EA 57.9 SGD
ALFAH50032.03 ALFAH50032.06
4-Aminonicotinic acid 98%
4-Aminonicotinic acid

Purity: 98% CAS No.: 7418-65-7 Molecular Formula: C6H6N2O2 Formula Weight: 138.13 Melting Point: 300° MDL No.: MFCD00234183

Formula: C₆H₆N₂O₂
MW: 138,13 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00234183
CAS Number: 7418-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR