5,6-Dichloronicotinic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
5,6-Dichloro-3-pyridinecarboxylic acid, 5,6-Dichloropyridine-3-carboxylic acid, 5,6-Dichloropyridine-3-carboxylicacid
B25586.06
B25586.14
ALFAB25586.06EA
0
SGD
ALFAB25586.06
ALFAB25586.14
5,6-Dichloronicotinic acid ≥98%
5,6-Dichloronicotinic acid
Purity: 98+%
CAS No.: 41667-95-2
Molecular Formula: C6H3Cl2NO2
Formula Weight: 192.00
Melting Point: ca 164° dec.
Beilstein Reference: 383740
Formula:
C₆H₃Cl₂NO₂ MW: 192 g/mol Melting Pt: ∼ 164 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00075181 CAS Number: 41667-95-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...