Hexane (mixture of isomers) ≥60,0% hexane_n 99,9 ACS organic_synthesis_LC_prep, General Laboratory Use

Supplier: BURDICK JACKSON
Danger

Synonyms: Hexanes, Isohexane

LP211-4 LP211D19
BJAALP211-4EA 2040 SGD
BJAALP211-4 BJAALP211D19
Hexane (mixture of isomers) ≥60,0% hexane_n 99,9 ACS organic_synthesis_LC_prep, General Laboratory Use
Hexane (mixture of isomers)

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 7.7 %(V)
Melting Pt: –95 °C
Density: 0.659…0.673 g/cm³ (20 °C)
Flash Pt: –26 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Passes test
Assay by GC Analysis (n-Hexane) 60 % min.
Assay by GC Analysis (total C6 isomers) 98.5 % min.
Color (APHA) 10 max.
Residue after Evaporation 0.001 % max.
Sulfur Compounds (as S) 0.005 % max.
Thiophene Passes test
Water by Karl Fischer Titration 0.01 % max.
Water-Soluble Titrable Acid 0.3 µeq/g max.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR