DURAN® Filter Discs with Hole, for Gas Distribution Tube
Supplier: DWK Life Sciences
254503008 254503102 254503205 254523001 254523104 254523207 254530108
SCOT254503008EA 94 SGD
SCOT254503008 SCOT254503102 SCOT254503205 SCOT254523001 SCOT254523104 SCOT254523207 SCOT254530108
DURAN® Filter Discs with Hole, for Gas Distribution Tube
Filters Filter Discs
This glass filter units are also used for gas and liquid distribution. For reactions between gases and liquids, gas distribution pipes are used in which filter plates with holes are processed, e.g. for gas washing bottles.
  • Manufactured from borosilicate glass 3.3
  • For gas distribution pipes
  • Available in porosities 0, 1 and 2
  • Glass articles for further processing