(2,4,6-Trimethylphenyl)acetonitrile ≥98%
Supplier: Thermo Scientific
|
Danger
|
B20534.03
B20534.06
B20534.14
ALFAB20534.03EA
0
SGD
ALFAB20534.03
ALFAB20534.06
ALFAB20534.14
(2,4,6-Trimethylphenyl)acetonitrile ≥98%
(2,4,6-Trimethylphenyl)acetonitrile
Purity: 98%
CAS No.: 34688-71-6
Molecular Formula: C11H13N
Formula Weight: 159.23
Melting Point: 77-80°
Boiling Point: 120°/1mm
UN No.: 3439
MDL No.: MFCD00013815
Formula:
C₁₁H₁₃N Boiling Pt: 120 °C (1 mmHg) Melting Pt: 77…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013815 CAS Number: 34688-71-6 EINECS: 252-151-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...