(4-Hydroxyphenyl)acetonitrile ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Hydroxybenzyl cyanide, 2-(4-Hydroxyphenyl)acetonitrile

A10971.03 A10971.06 A10971.14
ALFAA10971.03EA 0 SGD
ALFAA10971.03 ALFAA10971.06 ALFAA10971.14
(4-Hydroxyphenyl)acetonitrile ≥97%
(4-Hydroxyphenyl)acetonitrile

Purity: 97% CAS No.: 14191-95-8 Molecular Formula: C8H7NO Formula Weight: 133.15 Melting Point: 66-71° Boiling Point: 329-331° MDL No.: MFCD00002383

Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 329…331 °C
Melting Pt: 66…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00002383
CAS Number: 14191-95-8
EINECS: 238-046-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR