2-Amino-6-fluorobenzonitrile 99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Cyano-3-fluoroaniline

A12456.03 A12456.06 A12456.14
ALFAA12456.03EA 0 SGD
ALFAA12456.03 ALFAA12456.06 ALFAA12456.14
2-Amino-6-fluorobenzonitrile 99%
2-Amino-6-fluorobenzonitrile

Purity: 99% CAS No.: 77326-36-4 Molecular Formula: C7H5FN2 Formula Weight: 136.13 Melting Point: 125-129° Beilstein Reference: 3588945 UN No.: 3439 MDL No.: MFCD00015477

Formula: C₇H₅FN₂
MW: 136.13
Boiling Pt: 272
Melting Pt: 125…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00015477
CAS Number: 77326-36-4
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR