4-Amino-3-fluorobenzonitrile 99%

Supplier: Thermo Fisher Scientific
Danger

H26170.03 H26170.06
ALFAH26170.03EA 0 SGD
ALFAH26170.03 ALFAH26170.06
4-Amino-3-fluorobenzonitrile 99%
4-Amino-3-fluorobenzonitrile

Purity: 99% CAS No.: 63069-50-1 Molecular Formula: C7H5FN2 Formula Weight: 136.13 Melting Point: 87-90° UN No.: 3439 MDL No.: MFCD00055559

Formula: C₇H₅FN₂
MW: 136.13
Melting Pt: 87…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00055559
CAS Number: 63069-50-1
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR