2-Fluoro-5-iodobenzonitrile ≥97%

Supplier: Thermo Scientific
Danger

H26172.03 H26172.06
ALFAH26172.03EA 0 SGD
ALFAH26172.03 ALFAH26172.06
2-Fluoro-5-iodobenzonitrile ≥97%
2-Fluoro-5-iodobenzonitrile

Purity: 97% CAS No.: 351003-36-6 Molecular Formula: C7H3FIN Formula Weight: 247.01 Melting Point: 68-72° UN No.: 3439 MDL No.: MFCD03094225

Formula: C₇H₃FIN
MW: 247,01 g/mol
Melting Pt: 68…72 °C
Storage Temperature: Ambient
MDL Number: MFCD03094225
CAS Number: 351003-36-6
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR