5-Nitro-o-toluonitrile ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 2-Methyl-5-nitrobenzonitrile

A11392.06 A11392.14
ALFAA11392.06EA 0 SGD
ALFAA11392.06 ALFAA11392.14
5-Nitro-o-toluonitrile ≥98%
5-Nitro-o-toluonitrile

Purity: 98% CAS No.: 939-83-3 Molecular Formula: C8H6N2O2 Formula Weight: 162.15 Melting Point: 104-108° Beilstein Reference: 1947376 UN No.: 3439 MDL No.: MFCD00017040

Formula: C₈H₆N₂O₂
MW: 162,15 g/mol
Boiling Pt: 174 °C (18 mmHg)
Melting Pt: 104…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00017040
CAS Number: 939-83-3
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR