2-Fluoro-3-pyridineboronic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Fluoro-3-pyridinylboronic acid, (2-Fluoropyridin-3-yl)boronic acid, 2-Fluoropyridine-3-boronic acid
L20108.03
L20108.06
ALFAL20108.03EA
0
SGD
ALFAL20108.03
ALFAL20108.06
2-Fluoro-3-pyridineboronic acid ≥97%
2-Fluoro-3-pyridineboronic acid
Purity: 97%
CAS No.: 174669-73-9
Molecular Formula: C5H5BFNO2
Formula Weight: 140.91
Melting Point: 172°
Beilstein Reference: 9119935
Formula:
C₅H₅BFNO₂ MW: 140,91 g/mol Melting Pt: 172 °C Storage Temperature: Ambient |
MDL Number:
MFCD03411557 CAS Number: 174669-73-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...