3-Pyridineboronic acid

Supplier: Thermo Scientific
Warning

L15040.03 L15040.06
ALFAL15040.03EA 217 SGD
ALFAL15040.03 ALFAL15040.06
3-Pyridineboronic acid
3-Pyridineboronic acid

CAS No.: 1692-25-7 Molecular Formula: C5H6BNO2 Formula Weight: 122.92 Storage Temperature: 2°C to 8°C Melting Point: 300° Beilstein Reference: 471943

Formula: C₅H₆BNO₂
MW: 122,92 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00674177
CAS Number: 1692-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR