Water, B&J Brand™ for HPLC, for spectrophotometry, Burdick & Jackson™

Supplier: BURDICK JACKSON

Synonyms: Hydrogen oxide

NP365-10 NP365-20
BJAANP365-10EA 1460 SGD
BJAANP365-10 BJAANP365-20
Water, B&J Brand™ for HPLC, for spectrophotometry, Burdick & Jackson™
Water

  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 0.9971 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Fluorescence (as quinine base) 0.30 ppb max.
UV Absorbance @ 400 nm 0.005 AU max.
HPLC Gradient @ 200 nm 0.005 AU max.
Microbial Contamination Not detected
Refractive Index (20ºC) 1.3320 - 1.3440
Residue 1 mg/L max.
UV Absorbance @ 190 nm 0.01 AU max.
UV Absorbance @ 200 nm 0.01 AU max.
UV Absorbance @ 250 nm 0.005 AU max.
UV Absorbance @ 300 nm 0.005 AU max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR