3-Fluorostyrene 97% stabilised
Supplier: Thermo Scientific
|
Warning
|
Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC) 0,10 %
A12406.03
A12406.06
ALFAA12406.03EA
0
SGD
ALFAA12406.03
ALFAA12406.06
3-Fluorostyrene 97% stabilised
3-Fluorostyrene
Stabilized with 0.1% 4-tert-butylcatechol
Purity: 97%
CAS No.: 350-51-6
Molecular Formula: C8H7F
Formula Weight: 122.14
Storage Temperature: 2°C to 8°C
Flash Point: 29°(84°F)
Refractive Index: 1.5170
Odour: Recognizable
Boiling Point: 40-42°/8mm
Beilstein Reference: 2038488
UN No.: 1993
MDL No.: MFCD00000341
Formula:
C₈H₇F MW: 122,14 g/mol Boiling Pt: 40…42 °C (8 mmHg) Density: 1,025 Flash Pt: 29 °C (84 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000341 CAS Number: 350-51-6 EINECS: 206-504-9 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...