VWR® Sanitising Footbath Mats
Supplier: VWR Products
NP222.12X24X32 NP222.2.5X32X3
VWRMNP222.12X24X32EA 159 SGD
VWRMNP222.12X24X32 VWRMNP222.2.5X32X3
VWR® Sanitising Footbath Mats
Floor Mats Adhesive Floor Mats
Mats keep hygienic areas free of contamination and germs
  • Keeps hygienic areas free of contamination and germs
  • Thousands of pliable rubber scrapers dislodge contaminants from footwear as soles are immersed in disinfectant solution
  • Ideal for food-processing plants
The 61×81,3 cm mat is 1,3 cm thick and holds 3,8 L (1 gal.) of chlorine or quaternary disinfectant solution. The 81,3×99 cm tall-wall mat has 6,4 cm sides that allow it to hold up to 20,8 L (5,5 gal.) of sanitising solution. When used to fill the mat, the solution will totally immerse theMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable