Cable Ties, Inside Serrated
Supplier: HELLERMANN TYTON
111-00732 111-00718 111-00659
HELT111-00732EA 0 SGD
HELT111-00732 HELT111-00718 HELT111-00659
Cable Ties, Inside Serrated
Ties
E/TFE or Tefzel® cable ties are used when higher chemical resistance and/or temperatures up to 170 °C are required. These ties are most likely chosen for challenging applications in industries like food and beverage, aerospace, automotive or railway.
  • Blue cable tie offering high chemical resistance
  • Resistant to radioactivity and UV light
  • E/TFE is a non hydroscopic material, no moisture absorption
  • Complying with UL94 V0 requirements
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable