VWR® Cleanroom Latex Gloves, Class 10, Non Sterile
Supplier: VWR Collection
76579-672 76579-674 76579-676 76579-678
VWRU76579-672EA 415 SGD
VWRU76579-672 VWRU76579-674 VWRU76579-676 VWRU76579-678
VWR® Cleanroom Latex Gloves, Class 10, Non Sterile
Gloves
Ambidextrous, powder-free gloves designed to provide comfort and tactile sensitivity.
  • Ambidextrous, beaded cuff
  • Powder-free, silicone-free
  • Natural rubber latex
  • Low particle count
  • Specialised manufacturing process
Latex, nonsterile cleanroom gloves are available in Class 10 designation.
New Product
Order Now
SPECIFICATIONS
Ambidextrous / hand-specific Ambidextrous
Environmentally Preferable
Disposable / reusable Disposable
Latex / latex-free Latex
Powdered / powder-free Powder-free
Cuff style Beaded
Sterility Non sterile