Tetrahydrothiophen-3-one ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Dihydrothiophen-3(2H)-one, 4,5-Dihydrothiophen-3(2H)-one, 4,5-Dihydro-3(2H)-thiophenone
A11002.06
A11002.09
A11002.14
ALFAA11002.06EA
89.9
SGD
ALFAA11002.06
ALFAA11002.09
ALFAA11002.14
Tetrahydrothiophen-3-one ≥97%
Tetrahydrothiophen-3-one
Purity: 97%
CAS No.: 1003-04-9
Molecular Formula: C4H6OS
Formula Weight: 102.16
Flash Point: 77°(170°F)
Refractive Index: 1.5290
Odour: Characteristic
Boiling Point: 174-175°
Beilstein Reference: 105201
MDL No.: MFCD00005412
Formula:
C₄H₆OS MW: 102,16 g/mol Boiling Pt: 174…175 °C Density: 1,192 Flash Pt: 77 °C (170 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005412 CAS Number: 1003-04-9 EINECS: 213-698-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...