Aluminium lanthanum (III) oxide substrate 10x10x1 mm, polished one side 100 orientation

Supplier: Thermo Scientific

Synonyms: Lanthanum(III) aluminum oxide, Lanthanum Aluminium oxide, Aluminium lanthanum trioxide

38495.KT 38495.KU
ALFA38495.KTEA 483 SGD
ALFA38495.KT ALFA38495.KU
Aluminium lanthanum (III) oxide substrate 10x10x1 mm, polished one side 100 orientation
Aluminium lanthanum (III) oxide

CAS No.: 12003-65-5 Molecular Formula: LaAlO3 Odour: Odourless MDL No.: MFCD00144550

Formula: AlLaO₃
MW: 213,89 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00144550
CAS Number: 12003-65-5
EINECS: 234-433-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR