Heptane (mixture of isomers) ≥99.0%, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™

Supplier: BURDICK & JACKSON
Danger

PP206-19 PP206-204
BJAAPP206-19EA 1890 SGD
BJAAPP206-19 BJAAPP206-204
Heptane (mixture of isomers) ≥99.0%, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™
Heptane (mixture of isomers)

  • Low residue
  • Purified solvent
  • ACS General Use certified

Formula: H₃C(CH₂)₅CH₃
MW: 100,2 g/mol
Boiling Pt: 6,70 %(V)
Melting Pt: –90,6 °C
Density: 0,684 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dabde8793eb27ef796098b842bf18b9ced8539b8&vid=b57298f7f7280e366488444db9c9b34698895b06&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Passes test
GC Analysis (n-Heptane) 99 % min.
GC Analysis (total C7 isomers) 99.9 % min.
Refractive Index (20ºC) 1.3868 - 1.3888
Residue 5 mg/L max.
Water by Karl Fischer Titration 0.01 % max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR