Safety wash bottles, wide neck, vented, printed, Unitary™, Nalgene®
Supplier: Thermo Fisher Scientific
2436-0505
NALG2436-0505EA 54.3 SGD
NALG2436-0505
Safety wash bottles, wide neck, vented, printed, Unitary™, Nalgene®
Bottles Wash Bottles
LDPE, translucent with PP screw caps, except sodium hypochlorite (HDPE).
  • Good chemical resistance, heat resistant up to +80 °C
  • Ventilation system keeps hazardous volatile liquids in the bottle
  • Contents label with the chemical name, ICS chemical formula and CAS number
  • GHS compliant symbols
With hazard symbols and descriptions of hazards as required by the Hazardous Substance Ordinance. To allow easy hazard and chemical identification, and to prevent cross contamination, each bottle has a colour bar that corresponds to its cap colour. The design with no outlet tube and with laterally embedded spray tube prevents contamination when filling. The contents can be discharged by pressing without tipping or shaking the bottle. Colour coded caps with wide neck for easy filling.