5'-Chloro-2'-hydroxyacetophenone ≥99%

Supplier: Thermo Scientific
Warning

Synonyms: 1-(5-Chloro-2-hydroxyphenyl)ethan-1-one, 1-(5-Chloro-2-hydroxyphenyl)ethanone

A15448.03 A15448.06
ALFAA15448.03EA 0 SGD
ALFAA15448.03 ALFAA15448.06
5'-Chloro-2'-hydroxyacetophenone ≥99%
5'-Chloro-2'-hydroxyacetophenone

Purity: 99% CAS No.: 1450-74-4 Molecular Formula: C8H7ClO2 Formula Weight: 170.60 Melting Point: 52-53° Boiling Point: 107-109°/12mm MDL No.: MFCD00067788

Formula: C₈H₇ClO₂
MW: 170,6 g/mol
Boiling Pt: 107…109 °C (12 mmHg)
Melting Pt: 52…53 °C
Storage Temperature: Ambient
MDL Number: MFCD00067788
CAS Number: 1450-74-4
EINECS: 215-916-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR