Praseodymium ≥99.5% (REO, rare earth oxide basis), foil, Thickness: 0.62 mm (0.024 in)

Supplier: Thermo Scientific

00607.FF 00607.FI
ALFA00607.FFEA 740 SGD
ALFA00607.FF ALFA00607.FI
Praseodymium ≥99.5% (REO, rare earth oxide basis), foil, Thickness: 0.62 mm (0.024 in)
Praseodymium

Purity: 99.5% CAS No.: 7440-10-0 Physical Form: ≈2.6g/25x25mm, Packaged in mylar under argon Appearance: Grey Odour: Odourless MDL No.: MFCD00011174

Formula: Pr
MW: 140,91 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011174
CAS Number: 7440-10-0
EINECS: 231-120-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR