Copper ≥99.97% (metals basis), thin foil, Thickness: 0.015 mm (0.0006 in)

Supplier: Thermo Scientific

41504.FF 41504.FI
ALFA41504.FFEA 0 SGD
ALFA41504.FF ALFA41504.FI
Copper ≥99.97% (metals basis), thin foil, Thickness: 0.015 mm (0.0006 in)
Copper

Purity: 99.97% CAS No.: 7440-50-8 Physical Form: ≈0.084g/25x25mm, not certified pinhole free Odour: Odourless MDL No.: MFCD00010965

Formula: Cu
MW: 63,55 g/mol
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8,94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR