Chemical resistant gloves, KLEENGUARD* G80 Purple Nitrile
Supplier: KIMBERLY CLARK
97316 97317
KIMB97316EA 0 SGD
KIMB97316 KIMB97317
Chemical resistant gloves, KLEENGUARD* G80 Purple Nitrile
Gloves
The G80 gloves with the unique PURPLE NITRILE* formulation offer excellent chemical resistance against oils, greases, acids, caustics and solvents as well as durability. Made of 100% nitrile, they contain no natural rubber latex and thus reduce the potential for Type 1 glove associated reactions. Thanks to their ergonomic design these gloves provide maximum comfort and minimise hand fatigue.
  • Flock lined for extra comfort and easy donning
  • Textured palm and fingertips provide excellent grip in wet and dry situations
  • High abrasion and good puncture resistance, without compromising dexterity
Length: 325 mm
Thickness: 0,26 mm (cuff), 0,34 mm (finger)
Colour: Purple
EN 374 (AKL), EN 388 (4012), EN 374-2 (Level 2), AQL 1.5