(±)-1-Indanol ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(±)-1-Hydroxyindan, 1-Indanol, 2,3-Dihydro-1H-inden-1-ol
B24536.06
B24536.14
B24536.18
ALFAB24536.06EA
0
SGD
ALFAB24536.06
ALFAB24536.14
ALFAB24536.18
(±)-1-Indanol ≥98%
(±)-1-Indanol
Purity: 98%
CAS No.: 6351-10-6
Molecular Formula: C9H10O
Formula Weight: 134.18
Flash Point: 145°(293°F)
Melting Point: 50-54°
Boiling Point: 128°/12mm
Beilstein Reference: 2042960
MDL No.: MFCD00003797
Formula:
C₉H₁₀O MW: 134,18 g/mol Boiling Pt: 128 °C (12 mmHg) Melting Pt: 50…54 °C Flash Pt: 145 °C (293 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003797 CAS Number: 6351-10-6 EINECS: 224-230-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...