L(-)-Camphor ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: (-)-CAMPHOR, (1S,4S)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one

B23469.06 B23469.14
ALFAB23469.06EA 0 SGD
ALFAB23469.06 ALFAB23469.14
L(-)-Camphor ≥98%
L(-)-Camphor

Purity: 98% CAS No.: 464-48-2 Molecular Formula: C10H16O Formula Weight: 152.24 Flash Point: 65°(149°F) Odour: Camphor-like Boiling Point: 204° Beilstein Reference: 4291747 Merck Reference: 14,1732 Optical Rotation: -43° (c=10 in ethanol) UN No.: 2717 MDL No.: MFCD00064148

Formula: C₁₀H₁₆O
MW: 152,24 g/mol
Boiling Pt: 204 °C
Melting Pt: 175…180 °C
Density: 0,990
Flash Pt: 65 °C (149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064148
CAS Number: 464-48-2
EINECS: 207-354-7
UN: 2717
ADR: 4.1,III
Merck Index: 14,1732

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR