1-Bromo-6-chlorohexane ≥97%

Supplier: Thermo Fisher Scientific
Warning

B21619.06 B21619.14
ALFAB21619.06EA 0 SGD
ALFAB21619.06 ALFAB21619.14
1-Bromo-6-chlorohexane ≥97%
1-Bromo-6-chlorohexane

Purity: 97% CAS No.: 6294-17-3 Molecular Formula: Br(CH2)6ClFormula Weight: 199.53 Flash Point: 101°(214°F) Refractive Index: 1.4806 Boiling Point: 109-110°/2mm MDL No.: MFCD00001019

Formula: BrH₂C(CH₂)₄CH₂Cl
MW: 199.52 g/mol
Boiling Pt: 109…110 °C (2 mmHg)
Density: 1.337
Flash Pt: 101 °C (214 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001019
CAS Number: 6294-17-3
EINECS: 228-555-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR