(Methylthio)acetic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(Methylthio)ethansäure, 2-(Methylthio)acetic acid
B22531.14
B22531.03
B22531.06
ALFAB22531.14EA
628
SGD
ALFAB22531.14
ALFAB22531.03
ALFAB22531.06
(Methylthio)acetic acid ≥98%
(Methylthio)acetic acid
Purity: 98%
CAS No.: 2444-37-3
Molecular Formula: CH3SCH2CO2H
Formula Weight: 106.15
Flash Point:
Formula:
C₃H₆O₂S MW: 106,15 g/mol Boiling Pt: 99…100 °C (4 mmHg) Melting Pt: 13…14 °C Density: 1,224 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00075444 CAS Number: 2444-37-3 EINECS: 219-483-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...