N-Isopropyl-4-nitroaniline ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: N-isopropyl-4-nitroaniline

H52375.MD H52375.03 H52375.06
ALFAH52375.MDEA 67.9 SGD
ALFAH52375.MD ALFAH52375.03 ALFAH52375.06
N-Isopropyl-4-nitroaniline ≥97%
N-Isopropyl-4-nitroaniline

Purity: 97+% CAS No.: 25186-43-0 Molecular Formula: C9H12N2O2 Formula Weight: 180.20 UN No.: 2811 MDL No.: MFCD00128997

Formula: C₉H₁₂N₂O₂
MW: 180,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00128997
CAS Number: 25186-43-0
EINECS: 246-721-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR