1,8-Nonadiene ≥99%

Supplier: Thermo Scientific
Warning

Synonyms: Nona-1,8-diene

H53502.03 H53502.06 H53502.14
ALFAH53502.03EA 49.7 SGD
ALFAH53502.03 ALFAH53502.06 ALFAH53502.14
1,8-Nonadiene ≥99%
1,8-Nonadiene
Formula: C₉H₁₆
MW: 124,23 g/mol
Boiling Pt: 141…142 °C
Density: 0,74
Flash Pt: 26 °C (79 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008669
CAS Number: 4900-30-5
EINECS: 225-527-5
UN: 3295
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR