3-(Difluoromethoxy)aniline ≥97%

Supplier: Thermo Scientific
Danger

H53468.03 H53468.06
ALFAH53468.03EA 91.1 SGD
ALFAH53468.03 ALFAH53468.06
3-(Difluoromethoxy)aniline ≥97%
3-(Difluoromethoxy)aniline

Purity: 97% CAS No.: 22236-08-4 Molecular Formula: C7H7F2NO Formula Weight: 159.14 Refractive Index: 1.5100 Boiling Point: 201-202° MDL No.: MFCD00236221

Formula: C₇H₇F₂NO
MW: 159,14 g/mol
Boiling Pt: 201…202 °C
Density: 1,276
Storage Temperature: Ambient
MDL Number: MFCD00236221
CAS Number: 22236-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR