4-Bromo-2-fluorobenzyl chloride ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 4-Bromo-1-(chloromethyl)-2-fluorobenzene

L18680.06 L18680.03
ALFAL18680.06EA 0 SGD
ALFAL18680.06 ALFAL18680.03
4-Bromo-2-fluorobenzyl chloride ≥97%
4-Bromo-2-fluorobenzyl chloride

Purity: 97% CAS No.: 85510-82-3 Molecular Formula: C7H5BrClF Formula Weight: 223.47 Refractive Index: 1.5620 Beilstein Reference: 6323063 UN No.: 3265 MDL No.: MFCD02094036

Formula: C₇H₅BrClF
MW: 223,47 g/mol
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 37 °C
Density: 1,65
Storage Temperature: Ambient
MDL Number: MFCD02094036
CAS Number: 85510-82-3
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR