2-Chloro-3-(trifluoromethyl)phenol ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-Chloro-3-hydroxybenzotrifluoride
L19323.03
L19323.06
ALFAL19323.03EA
0
SGD
ALFAL19323.03
ALFAL19323.06
2-Chloro-3-(trifluoromethyl)phenol ≥98%
2-Chloro-3-(trifluoromethyl)phenol
Purity: 98%
CAS No.: 138377-34-1
Molecular Formula: C7H4ClF3O
Formula Weight: 196.56
Flash Point: 79°(174°F)
Refractive Index: 1.4730
Boiling Point: 160°
Beilstein Reference: 9045773
UN No.: 3265
MDL No.: MFCD00213793
Formula:
C₇H₄ClF₃O MW: 196,56 g/mol Boiling Pt: 160 °C Density: 1,479 Flash Pt: 79 °C (174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00213793 CAS Number: 138377-34-1 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...