1-Chloro-2-octyne ≥98%
Supplier: Thermo Scientific
|
Warning
|
H53452.06
H53452.14
ALFAH53452.06EA
131
SGD
ALFAH53452.06
ALFAH53452.14
1-Chloro-2-octyne ≥98%
1-Chloro-2-octyne
Purity: 98%
CAS No.: 51575-83-8
Molecular Formula: C8H13Cl
Formula Weight: 144.64
Flash Point: 68°(154°F)
Refractive Index: 1.46
Boiling Point: 81°/15mm
MDL No.: MFCD00274223
Formula:
C₈H₁₃Cl MW: 144,64 g/mol Boiling Pt: 81 °C (15 mmHg) Density: 0,931 Flash Pt: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00274223 CAS Number: 51575-83-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...