5-Chloro-2-methoxybenzonitrile ≥98%
Supplier: Thermo Scientific
|
Danger
|
A15185.03
A15185.06
ALFAA15185.03EA
0
SGD
ALFAA15185.03
ALFAA15185.06
5-Chloro-2-methoxybenzonitrile ≥98%
5-Chloro-2-methoxybenzonitrile
Purity: 98%
CAS No.: 55877-79-7
Molecular Formula: C8H6ClNO
Formula Weight: 167.60
Melting Point: 98-102°
Beilstein Reference: 3245801
UN No.: 3439
MDL No.: MFCD00052867
Formula:
C₈H₆ClNO MW: 167,59 g/mol Boiling Pt: 276 °C (1013 hPa) Melting Pt: 98…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052867 CAS Number: 55877-79-7 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...