4'-Chloro-3'-nitroacetophenone ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Chloro-3-nitroacetophenone

A15121.06 A15121.14
ALFAA15121.06EA 0 SGD
ALFAA15121.06 ALFAA15121.14
4'-Chloro-3'-nitroacetophenone ≥97%
4'-Chloro-3'-nitroacetophenone

Purity: 97% CAS No.: 5465-65-6 Molecular Formula: C8H6ClNO3 Formula Weight: 199.60 Melting Point: 96-100° Beilstein Reference: 1640627

Formula: C₈H₆ClNO₃
MW: 199,59 g/mol
Melting Pt: 96…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00007083
CAS Number: 5465-65-6
EINECS: 226-769-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR