2-Fluoro-5-methoxybenzonitrile ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 1-Cyan-2-fluor-5-methoxybenzene

H32724.03 H32724.06
ALFAH32724.03EA 0 SGD
ALFAH32724.03 ALFAH32724.06
2-Fluoro-5-methoxybenzonitrile ≥98%
2-Fluoro-5-methoxybenzonitrile

Purity: 98% CAS No.: 127667-01-0 Molecular Formula: C8H6FNO Formula Weight: 151.14 Melting Point: 81-85° UN No.: 3439 MDL No.: MFCD00142921

Formula: C₈H₆FNO
MW: 151,14 g/mol
Melting Pt: 81…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00142921
CAS Number: 127667-01-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR