3-Fluoro-4-iodotoluene ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Fluoro-1-iodo-4-methylbenzene, 1-Fluoro-2-iodo-5-methylbenzene
H53491.03
H53491.06
ALFAH53491.03EA
134
SGD
ALFAH53491.03
ALFAH53491.06
3-Fluoro-4-iodotoluene ≥97%
3-Fluoro-4-iodotoluene
Purity: 97%
CAS No.: 452-79-9
Molecular Formula: C7H6FI
Formula Weight: 236.03
Flash Point: 92°(198°F)
Refractive Index: 1.5810
Boiling Point: 138-139°/9mm
MDL No.: MFCD00143162
Formula:
C₇H₆FI MW: 236,03 g/mol Boiling Pt: 138…139 °C (9 mmHg) Density: 1,803 Flash Pt: 92 °C (198 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00143162 CAS Number: 452-79-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...